Masterflex® Ismatec® Quick-Couple, Single-Channel Pump Heads for 1.6 or 2.4 mm Wall Tubing, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX78002-11EA 1380 EUR
MFLX78002-11 MFLX78002-16 MFLX78002-22
Masterflex® Ismatec® Quick-Couple, Single-Channel Pump Heads for 1.6 or 2.4 mm Wall Tubing, Avantor®
Pumput Pumppupäät Peristalttiset pumppupäät
Snap your pump head onto your drives quickly with turn-and-lock mounting system.
  • Select from interchangeable pump heads that offer flow rates from 0.01 to 3660 ml/min
Ensure optimum performance from your Ismatec® pumps.