Masterflex® Ismatec® Quick-Couple Pump Head for 1.6 mm Wall Tubing, 2 Rollers, 3700 mL/min
Toimittaja: Avantor Fluid Handling
VWR Item#MFLX78002-11
Your Organization's Price:
Unavailable
Snap your pump head onto your drives quickly with turn-and-lock mounting system.
  • Select from interchangeable pump heads that offer flow rates from 0.01 to 3660 ml/min
  • 2 rollers
Ensure optimum performance from your Ismatec® pumps.