Mobile concentrator system, CentriVap®
Toimittaja: LABCONCO
7812035 7812036
LBCP7812035EA 14200 EUR
LBCP7812035 LBCP7812036
Mobile concentrator system, CentriVap®
Haihduttimet Sentrifugaaliset haihduttimet
This mobile system with or without heat boost combines the separate vacuum concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to laboratory. A 12 to 13 mm rotor and solvent trap are also included. The interior can accommodate a vacuum pump — the only additional component necessary to begin processing.
  • Microprocessor-controlled centrifugal motion up to 1725 rpm eliminates bumping and foaming
  • Quick-Stop™ System:Once run time has expired or the RUN/STOP button is pressed, within seconds the