Centrifugal concentrators, complete systems and system packages, CentriVap®
Toimittaja: LABCONCO
7982011 7983014 7982035 7983035 7989601 7989635 7315032 7315030 7315033 7315031 7315062 7315060 7315061 7315063 7995811
531-0223EA 16020 EUR
531-0223 531-0224 LBCP7982035 LBCP7983035 535-0409 LBCP7989635 431-0235 431-0231 431-0237 431-0233 431-0236 431-0232 431-0234 431-0238 LBCP7995811
Centrifugal concentrators, complete systems and system packages, CentriVap®
Haihduttimet Sentrifugaaliset haihduttimet
CentriVap® vacuum concentrators are available as complete systems or system packages. The acid resistant model is recommended for corrosive solvents (TFA, acetic acid, and so on). Other models feature Heat Boost, which surrounds the chamber wall, to further speed evaporation of high boiling point solvents.
  • Epoxy-coated aluminium chamber
  • Centrifugation, controlled by microprocessor, up to 1725 min⁻¹ and heating, controlled by microprocessor up to 100 °C
  • Control panel with on/off button, pre-heating button, optional Heat Boost button, parameter setting,