RapidVap®-vakuumihaihdutusjärjestelmät
Toimittaja: LABCONCO
7900001 7900003 7900011 7900013
531-0171EA 19230 EUR
531-0171 531-0172 531-0219 531-0220
RapidVap®-vakuumihaihdutusjärjestelmät
Haihduttimet Sentrifugaaliset haihduttimet
These systems use vacuum along with heat and vortex motion to help speed evaporation and provide an alternative to traditional centrifugal evaporation methods. The systems may be used with a broad range of aggressive chemicals. Heat is supplied by a dry block heating system that, unlike water baths, adds no potential source of contamination and requires no maintenance.
  • Fast evaporation rates; water evaporates at a rate up to 0,75 ml/minute/tube
  • Microprocessor controlled 1000 W dry block heating system up to +100 °C
  • Microprocessor controlled