Salicylohydroxamic acid ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAS0008-5GEA 20.9 EUR
TCIAS0008-5G TCIAS0008-25G
Salicylohydroxamic acid ≥98.0% (by HPLC, titration analysis)
Salicylohydroxamic acid
Kaava: C₇H₇NO₃
MW: 153,14 g/mol
MDL numero: MFCD00002110
CAS-numero: 89-73-6
EINECS: 201-934-3
Merck Index: 14,08331
https://fi.vwr.com/stibo/web/std.lang.all/38/86/28993886.jpg

Order Now

Testitulosten erittely

Appearance White to Light red to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR