1-Fenyylipyratsoli ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAP1868-5GEA
41.8
EUR
TCIAP1868-5G
TCIAP1868-25G
1-Fenyylipyratsoli ≥98.0% (by GC)
1-Fenyylipyratsoli
Kaava:
C₉H₈N₂ MW: 144,18 g/mol Kiehumispiste: 142°C Sulamispiste: 11°C |
MDL numero:
MFCD00003112 CAS-numero: 1126-00-7 EINECS: 214-415-1 |
Testitulosten erittely
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...