(Acetylmethylene)triphenylphosphorane 99%
Toimittaja: Thermo Scientific
ACRO169970250EA
104
EUR
ACRO169970250
ACRO169971000
(Acetylmethylene)triphenylphosphorane 99%
(Acetylmethylene)triphenylphosphorane
Kaava:
C₂₁H₁₉OP MW: 318,36 g/mol Sulamispiste: 203…207 °C Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00008774 CAS-numero: 1439-36-7 EINECS: 215-878-2 |
Testitulosten erittely
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 203 to 211 °C |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...