VWR®, Jäähdyttävät termostaattihauteet, MX-lämpötilasäädin
Toimittaja: VWR
VWR®
462-0223EA 4380 EUR
462-0223 462-0225 VWRU89202-966 VWRU89202-968 VWRU89202-994
VWR®, Jäähdyttävät termostaattihauteet, MX-lämpötilasäädin
Kiertotermostaatit Suljetut termostaattihaudejärjestelmät Jäähdyttävät termostaattihauteet
Stainless steel baths with user friendly MX temperature controller plus three control buttons and on-screen prompts. Baths can be used for external closed loop circulation. The controller rests on a chemically resistant DuraTop™ deck, which is cool to the touch, even when working at high temperatures, and units feature LidDock™ lid stowing system. Cool Command™ technology regulates the amount of cooling required, saving energy while providing rapid cooling and precise control at elevated temperatures.
  • Large EasyView™ LCD display with alarm or fault icons plus English prompts and menus
  • User-adjustable high and low temperature limits and low liquid level alarms
  • Single speed pressure pump with external closed loop circulation capability
  • Integral pump and heater coil protection
  • Single point calibration
Maximum pump capacities:
11,9 L/min, 120 mbar.
Sertifikaatit: Complies with DIN 12876-1, Safety Class I.
Toimitustiedot: Supplied with a reservoir cover, bypass tubing, male inlet and outlet adapters for 47, 63 and 95 mm tubing, ¼" to M16 adapters are also included.
Varoitus: Cooling capacity at 20 °C