Ruiskuttomat suodattimet, Mini-UniPrep™, Whatman™
Toimittaja: Whatman products (Cytiva)
Whatman™ Mini-UniPrep™
514-8119EA 390 EUR
514-8119 514-8118 514-8117 514-8116 514-8115 514-8114 514-8113 514-8079 514-8112 514-8111 514-8078 514-8077 514-8110 514-8076 514-8075 516-6355 516-6356 516-6343 514-8109 561-6341 514-8108 514-8107 514-8105 514-8104 514-8103 514-8102 514-8100 516-0993 514-8099 514-8131 514-8098 514-8097 514-8096 514-8095 514-8094 514-8093 514-8092 514-8091 514-8090 514-0124 514-0125 514-0120 514-8129 514-0123 514-8127 514-8126 514-8125 514-8124 514-8123 514-8089 514-8122 514-8121 514-8120 514-8086 514-8085 514-8084 514-8083 514-8082 514-8080
Ruiskuttomat suodattimet, Mini-UniPrep™, Whatman™
Suodattimet Ruiskuttomat suodattimet
The Whatman Mini-UniPrep Syringeless Filters combine four products into one syringeless filter solution for efficient chromatography sample preparation for a broad range of applications. They are built for fast and easy high performance liquid chromatography/ultra high performance liquid chromatography sample preparation and analysis.
  • All-in-one filtration device for quick and cost-effective chromatography sample processing
  • Eliminates the need for separate syringes, syringe membrane filters, vials, and septa
  • Designed for use with an UHPLC or HPLC autosampler for high-throughput analysis
  • 0,2 µm and 0,45 µm filter pore sizes available to meet filtration sample requirements
  • Compatible with most major autosamplers
  • Housing options: amber to prevent photodegradation of light-sensitive samples, or translucent for easy visual inspection made from polypropylene (PP) material
  • Recyclable after use
By excluding large particles from a sample, the time required to process the mobile phase is shortened. Also, column life is extended by reducing contamination that ultimately causes clogging and back pressure build up.
Applications include: Routine HPLC/UHPLC analysis, composite assays, content uniformity, protein precipitation, solubility testing, dissolution testing and sample filtration. Use with any compound that requires light protection, such as catecholamines or vitamins.
These filters can be used either manually or with a compressor unit. Compressor units allow for processing of multiple samples, further reducing sample preparation time for chromatographic analysis.
PP elatusaine- tai liuotinpohjaisten näytteiden yleissuodatukseen.
PTFE: vahvoille kemikaaleille, kalvo erittäin kemikaalinkestävä
Regeneroitu selluloosa ja PVDF vesipohjaisille tai orgaanisille liuottimille - vähän proteiineja sitova kalvo.
Nailon: vesipohjaisille/orgaanisille näytteille pH-alueella 3 - 10.
PES: Biologisille näytteille, jotka edellyttävät vähän proteiineja sitovaa suodatinkalvoa.
PP: vesipohjaisille/orgaanisille liuottimille, suuri virtausnopeus, kestävä kalvomateriaali.
GMF for high particulate laden liquids.
Suodatuskapasiteetti: 0,4 ml
Enimmäiskäyttölämpötila: 50 °C