Neon lamps
Toimittaja: Phywe
765-1203EA 13.3 EUR
765-1203
Neon lamps
Valot Neonlamput
Tubular glow lamp of glass. With conical contact hats at the ends. Used, inter alia, to determine the polarity of charge, because light only occurs around the negative electrode. Ignition voltage approximately 80 V, switching voltage approximately 65 V. For example, connects with crocodile clips.
Length: 50 mm
Length: 50 mm
Ø: 10 mm