2-Amino-5-bromopyrimidine

Toimittaja: Molekula

MOLEM20340532EA 178 EUR
MOLEM20340532
2-Amino-5-bromopyrimidine
2-Amino-5-bromopyrimidine
Kaava: C₄H₄BrN₃
MW: 174 g/mol
MDL numero: MFCD00012341
CAS-numero: 7752-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR