Thermo Scientific™ Matrix™ Sample Storage Starter Packs
Toimittaja: Thermo Scientific
Matrix™
479-2466EA 2707.5 EUR
479-2466 479-2465 479-2468 479-2467 479-2464
Thermo Scientific™ Matrix™ Sample Storage Starter Packs
Näytevialit ja pienet näyteastiat
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs.
  • Eliminate the need to reformat non-2D cryotube samples
  • Secure sample tracking with 2D, 1D, and human readable codes
  • Scan 96 tubes in as little as 1 second with advanced and premium starter packs
  • Help ensure quality with ISO 7 class cleanroom - manufactured tubes with sterility assurance
Choose from five different starter pack options.