Anti-HCP5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Toimittaja: Bioss
BOSSBS-15429R-A750EA 855 EUR
BOSSBS-15429R-A750
Anti-HCP5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Vasta-aineet
HCP5 is a region present on chromosome 6p21.3 that is characterised by multiple duplicated gene families. HCP5 (HLA class I histocompatibility antigen protein P5), also known as P5-1, is a 132 amino acid protein that is encoded by a gene mapping to human chromosome 6p21.33. Localising within the MHC class I region, HCP5 is not structurally related to other MHC class I genes, but does have high sequence similarity with HERV-L and HERV-16. HCP5 also has high sequence homology to retroviral Pol genes, making it a possible candidate for interaction with HIV-1 through an antisense mechanism that prevents retrovirus transcription. Single-nucleotide polymorphisms (SNPs) to the region of the HCP5 gene that corresponds with HLA-B has been linked to a lower HIV-1 viral set point. HCP5 is highly expressed in lymphoid tissues, spleen and activated lymphocytes, as well as B-cell and natural killer (NK) cell lines.

Type: Primary
Antigen: HCP5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-80/132
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR