Anti-HCA25a Rabbit Polyclonal Antibody (FITC)

Toimittaja: Bioss
BOSSBS-15421R-FITCEA 858 EUR
BOSSBS-15421R-FITC
Anti-HCA25a Rabbit Polyclonal Antibody (FITC)
Vasta-aineet
HCA25a.

Type: Primary
Antigen: HCA25a
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 1-100/153
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR