Anti-C9ORF140 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Toimittaja: Bioss
BOSSBS-15314R-A488EA 858 EUR
BOSSBS-15314R-A488
Anti-C9ORF140 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Vasta-aineet
C9orf140 (chromosome 9 open reading frame 140), also known as TS/MDEP (tumor specificity and mitosis phase-dependent expression protein) or p42.3, is a 394 amino acid nuclear and cytoplasmic protein encoded by a gene that maps to human chromosome 9q34.3. Chromosome 9 consists of about 145 million bases, represents 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterised by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias.

Type: Primary
Antigen: C9orf140
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 301-394/394
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR