Reliable hotplate with exceptional safety features for everyday use.
- SafetyHeat™ system protects the laboratory
- Easy to read LCD display with everything you need to know
- Designed for durability and reliability with SmartHousing™
OHAUS Guardian 5000™ hotplates are designed for safety and simplicity in all research, academic and industrial applications. The design ensures proper temperature control of the top plate or your sample when using the optional temperature probe.
These heating models include a Single Point Calibration feature for multiple setpoints. Ideal for everyday heating applications.
Tilaustiedot: Supplied without optional temperature probe which must be ordered separately.