Deuteriumoksidi
Toimittaja: TCI EUROPE
TCIAW0002-10MLEA
103
EUR
TCIAW0002-10ML
TCIAW0002-100ML
Deuteriumoksidi
Deuteriumoksidi
Kaava:
D₂O MW: 20 g/mol Kiehumispiste: 101°C Sulamispiste: 4°C |
MDL numero:
MFCD00044636 CAS-numero: 7789-20-0 EINECS: 232-148-9 |
Testitulosten erittely
Appearance | Colorless to Almost colorless clear liquid |
Degree of deuteration | min. 99.8 atom%D |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...