Methyl-2,3,4-tri-O-benzyl-1-thio-β-L-fucopyranoside ≥95.0% (by HPLC)

Toimittaja: TCI EUROPE

TCIAM1628-1GEA 433 EUR
TCIAM1628-1G
Methyl-2,3,4-tri-O-benzyl-1-thio-β-L-fucopyranoside ≥95.0% (by HPLC)
Methyl-2,3,4-tri-O-benzyl-1-thio-beta-L-fucopyranoside
Kaava: C₂₈H₃₂O₄S
MW: 464,62 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD06797141
CAS-numero: 107802-80-2

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 53.0 to 58.0 deg-C
Specific rotation [a]20/D 0.0 to +3.0 deg (C=1, CHCl3)
Solubility in Chloroform very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR