3,3'-Dimethylazobenzene ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAD0676-1GEA 68.2 EUR
TCIAD0676-1G TCIAD0676-5G
3,3'-Dimethylazobenzene ≥98.0% (by GC)
3,3'-Dimethylazobenzene
Kaava: C₁₄H₁₄N₂
MW: 210,28 g/mol
MDL numero: MFCD00048080
CAS-numero: 588-04-5
EINECS: 209-609-8

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 54.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR