ESD safety shoes, uvex motion style, 6978
Toimittaja: Uvex
uvex motion style
UVEX6978.839EA 146 EUR
UVEX6978.839 UVEX6978.843 UVEX6978.842 UVEX6978.845 UVEX6978.844 UVEX6978.847 UVEX6978.836 UVEX6978.846 UVEX6978.838 UVEX6978.837 UVEX6978.848 UVEX6978.841 UVEX6978.840
ESD safety shoes, uvex motion style, 6978
Kengät ja saappaat
Ultra-light ESD safety shoe made from breathable high-tech micro velour, soft padded collar and tongue, breathable distance mesh plus an extremely flexible dual-density PUR sole (slip, abrasion and cut resistant, resistant to oil and petrol also withstands brief exposure to heat of approx. +120 °C). Free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • Protective steel toecap (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/grey
Width: 11
Sertifikaatit: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S2, SRC.