Anti-IgA Human Polyclonal Antibody (Unconjugated)
Toimittaja: Immunoreagents
IMMRIR2367EA 136 EUR
IMMRIR2367
Anti-IgA Human Polyclonal Antibody (Unconjugated)
Vasta-aineet
Type:
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype:
Reactivity:
Order Now