Anti-IgG Goat Polyclonal Antibody (Biotin)

Toimittaja: Immunoreagents
IMMRGT-003-ZBIOEA 98.5 EUR
IMMRGT-003-ZBIO
Anti-IgG Goat Polyclonal Antibody (Biotin)
Vasta-aineet
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR