HT (hypoxanthine, thymidine) media, cellgro™, Corning®
Toimittaja: Corning
Corning®
MDTC25-047-CIEA 21.39 EUR
MDTC25-047-CI
HT (hypoxanthine, thymidine) media, cellgro™, Corning®
Soluviljelyyn liuokset
HT media is a 'rescue media' that provides pre-formed purines and pyrimidines to overcome the effects of residual intracellular aminopterin.
  • Complete Certificate of Analysis available for each production lot, along with full formulation information
  • Produced under the highest industry standards to ensure superior results