Anti-FCAR Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Toimittaja: Bioss
BOSSBS-6586R-A555EA 858 EUR
BOSSBS-6586R-A555
Anti-FCAR Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Vasta-aineet
Binds to the Fc region of immunoglobulins alpha. Mediates several functions including cytokine production. Tissue specificity; Isoform A.1, isoform A.2 and isoform A.3 are differentially expressed between blood and mucosal myeloid cells. Isoform A.1, isoform A.2 and isoform A.3 are expressed in monocytes. Isoform A.1 and isoform A.2 are expressed in alveolar macrophages; however only one isoform is expressed at alveolar macrophages surfaces.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FCAR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR