Anti-NGBR Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Toimittaja: Bioss
BOSSBS-11468R-A647EA 858 EUR
BOSSBS-11468R-A647
Anti-NGBR Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Vasta-aineet
Nogo is an oligodendrocyte-specific member of the Reticulon family and is a component of CNS white matter that inhibits axon outgrowth, induces collapse of growth cones of chick dorsal root ganglion cells, and inhibits the spreading of 3T3 fibroblasts. Nogo is expressed by oligodendrocytes but not by Schwann cells and associates primarily with the endoplasmic reticulum. Nogo exists in three different splice forms, Nogo-A, -B and -C. NgBR (Nogo-B receptor), also known as nuclear undecaprenyl pyrophosphate synthase 1 homolog, is a 293 amino acid single-pass type I membrane protein that acts as a specific receptor for the amino-terminus of Nogo-B. Through this interaction, NgBR is involved in the regulation of vascular remodeling and angiogenesis. NgBR also enhances Niemann-Pick type C2 protein (NPC2) stabilization. Knockdown of NgBR mRNA leads to decreased NPC2 levels, which results in the hallmarks of NPC2 mutation: increased intracellular cholesterol accumulation and a loss of sterol sensing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NGBR/Nogo B receptor
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR