Suojapuku, kemikaalinkestävä, HOP'SAFE EXTRA
Toimittaja: Kolmi-Hopen
HOPE40.070/XLEA 99.3 EUR
HOPE40.070/XL HOPE40.070/S HOPE40.070/L HOPE40.070/M HOPE40.070/XXL
Suojapuku, kemikaalinkestävä, HOP'SAFE EXTRA
Kertakäyttöiset vaatteet
These overalls made of breathable SMS fabric with microporous, waterproof PE membrane provide good protection against liquid aerosols and airborne solid particulate chemicals and limited protection against liquid mist. They do not only protect the wearer but also products and processes from contamination.
  • Elasticated hood, cuffs, ankles and waist
  • With zip closure with flap
  • External seams for comfort
  • Lightweight and flexible, permeable to air and water vapor
  • Good resistance to tears and abrasion
  • Electrostatic protection: 2,49×10⁹ Ω
Ideal for use in the chemical industry, petrochemistry, industrial maintenance and pharmaceutical industry.
Weight: 65 g/m²
Sertifikaatit: Type 4B/5B/6B, EN 1149-5, EN 1073-2, EN 14126; Cat. III
Pakkaus: Individually packed.