Anti-FBLIM1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Toimittaja: US Biological
035529-AP
USBI035529-APEA 884 EUR
USBI035529-AP
Anti-FBLIM1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: FBLIM1 (filamin binding LIM protein 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human