Anti-KIRREL2 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
ORB100369-100 ORB100369-200
BIRBORB100369-100EA 0 EUR
BIRBORB100369-100 BIRBORB100369-200
Anti-KIRREL2 Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: KIRREL2 (kin of IRRE like 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat