Culture media, ready to use, in plates, 90 mm
Toimittaja: bioMérieux Industry
AEB120102EA 83.5 EUR
AEB120102 AEB520710 BMERAEB522860 AEB520100 BMERAEB522360 AEB523510
Culture media, ready to use, in plates, 90 mm
Elatusaineet mikrobiologiaan Valmisteltu viljelyaine
With more than 50 year experience in microbiology, bioMérieux has developed a complete collection of culture media, especially designed for food sample testing and for the medical field.
A wide range of culture media, specifically devised and evaluated for food testing, is available to detect pathogens such as Listeria monocytogenes, Salmonella, E. coli O157:H7, Campylobacter, Vibrio and more.