SPE columns, Supra-Clean® SI-S (silica)
Toimittaja: PerkinElmer
Supra-Clean®
PERCN9306534EA 163 EUR
PERCN9306534 PERCN9306402 PERCN9306535 PERCN9306532 PERCN9306466 PERCN9306444 PERCN9306533 PERCN9306404
SPE columns, Supra-Clean® SI-S (silica)
Kromatografiakolonnit
SPE Supra-Clean® SI-S are spherical silica-based columns. Ideal for extracting polar compounds when analysing aldehydes, amines, pesticides, herbicides, carotenoids, fat soluble vitamins, baflatoxins, fatty acids, and phospholipids.