Hengityssuojaimen suodatin, X-plore® Rd90
Toimittaja: Dräger
X-plore®
DRAE6738870EA 217 EUR
DRAE6738870 DRAE6738865 DRAE6738866 111-1546 DRAE6736705
Hengityssuojaimen suodatin, X-plore® Rd90
Hengityssuojaimet Uudelleen käytettävät maskit Suojainten suodattimet
The filters of the Dräger X-plore® Rd90 series protect from hazardous gases and vapors, have a long life and can be used for many different masks.
  • Filters have been designed for use in the Dräger half masks, X-plore® 4390 and 4790. With the 40/90 filter box adapter they can also be used for any mask with a Rd40 connection in accordance with EN 148-1.
  • Gas filters and combination filters have a shelf-life of 6 years and, the particle filters a shelf life of 12 years from date of manufacture
  • Filters are packed individually to ensure optimum protection of the unused filter
  • Aluminium housing ensures that any damage to the filter is easily detected, therefore providing extra security
The filters are suitable for use with half and full face masks with standard thread connection Rd40 according to EN 148-1.
Sertifikaatit: EN 14387, EN 143, EN 148-1