Bromifenolisininen 0,04% in water for pH determination

Toimittaja: TCI EUROPE

TCIAB0117-500MLEA 35.2 EUR
TCIAB0117-500ML
Bromifenolisininen 0,04% in water for pH determination
Bromifenolisininen
Kaava: C₁₉H₁₀Br₄O₅S
MW: 669,97 g/mol
MDL numero: MFCD00005875
CAS-numero: 115-39-9

Order Now

Testitulosten erittely

Transition interval (pH) (yellow)3.0 to 4.6 (lavender)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR