Masterflex® Transfer Tubing, Cleargreen® PVC, 5/6" ID x 7/16" OD; 50 Ft
Toimittaja: Avantor Fluid Handling
VWR Item#MFLX96605-29
Your Organization's Price:
Eliminate odor transferring to sample.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 5/6" ID x 7/16" OD
<B>Characteristics:</B> Translucent, odorless, low water absorption, low extractables <br /><B>Certified compliant</B> FDA compliant, USP Class VI, 3A compliant, Reach compliant and RoHS compliant.<br /><B>Temperature range:</B> -58 to 165 °F (-50 to 74 °C) <br /><B>Sterilize:</B> Autoclave or ethylene oxide.