Masterflex® Transfer Tubing, Cleargreen® PVC, 3/16" ID x 5/16" OD; 50 Ft
Toimittaja: Avantor Fluid Handling
Eliminate odor transferring to sample.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Tubing size: 3/16" ID x 5/16" OD
<B>Characteristics:</B> Translucent, odorless, low water absorption, low extractables <br /><B>Certified compliant</B> FDA compliant, USP Class VI, 3A compliant, Reach compliant and RoHS compliant.<br /><B>Temperature range:</B> -58 to 165 °F (-50 to 74 °C) <br /><B>Sterilize:</B> Autoclave or ethylene oxide.