Overhead stirrer tools, propeller, diamond, centrifugal and anchor-type impellers
Toimittaja: witeg Labortechnik
DH.WOS501010 DH.WOS501013 DH.WOS501015 DH.WOS501017 DH.WOS501020 DH.WOS501031 DH.WOS501035 DH.WOS501111 DH.WOS501135
WITGDH.WOS501010EA 38 EUR
WITGDH.WOS501010 WITGDH.WOS501013 WITGDH.WOS501015 WITGDH.WOS501017 WITGDH.WOS501020 WITGDH.WOS501031 WITGDH.WOS501035 WITGDH.WOS501111 WITGDH.WOS501135
Overhead stirrer tools, propeller, diamond, centrifugal and anchor-type impellers
Sekoittajat Sekoitustyökalut
Stirring impeller for high speed-type and high torque-type overhead stirrer.
  • Made of stainless steel 304 (high-grade)
  • Resistant to acids, chemicals, corrosion and high temperature