Sekoitusmoottori, mekaaniset, WiseStir® HS-50A
Toimittaja: witeg Labortechnik
DH.WOS01210 DH.WOS01211
WITGDH.WOS01210EA 745 EUR
WITGDH.WOS01210 WITGDH.WOS01211
Sekoitusmoottori, mekaaniset, WiseStir® HS-50A
Sekoittajat Sekoitusmoottorit
General purpose basic laboratory overhead stirrer, ideal for stirring low to medium viscosity materials. The analogue unit has a chrome-plated stainless steel chuck to prevent corrosion. Ideal for use in schools, universities and inspection laboratories.
  • Available with smooth start, change of stirring speed
  • Easy to use dial for setting the speed
  • The position of the impeller is adjustable
  • Motor is protected with over heat function
Tilaustiedot: Supplied without accessories, a wide range of accessories are available to order separately.