Cell scrapers
Toimittaja: witeg Labortechnik
8.486.200
WITG8.486.200EA 158 EUR
WITG8.486.200
Cell scrapers
Soluraapat
Bent, plastic coated glass handle, mobile PTFE blade. Scraper for removal of cells from flasks and plates. Length 50 mm.