Safety wash bottles, narrow neck, with printed name
Toimittaja: witeg Labortechnik
7.169.109 7.169.110 7.169.111 7.169.113 7.169.114 7.169.116 7.169.142 7.169.143
WITG7.169.109EA 9.5 EUR
WITG7.169.109 WITG7.169.110 WITG7.169.111 WITG7.169.113 WITG7.169.114 WITG7.169.116 WITG7.169.142 WITG7.169.143
Safety wash bottles, narrow neck, with printed name
Pullot Pesupullot
LDPE, yellow, with coloured screw caps.
  • With hazard symbols and descriptions of hazards as required by the hazardous substance ordinance
  • Good chemical resistance
  • Good thermal resistance
With safety delivery jet.