Stopcocks for desiccators
Toimittaja: witeg Labortechnik
5.662.004
WITG5.662.004EA 85.4 EUR
WITG5.662.004
Stopcocks for desiccators
Hanat
Stopcock, with FCH-V 24/29, flat-free high vacuum joint, made of borosilicate glass.