Centrifuge tubes, round bottom
Toimittaja: witeg Labortechnik
4.940.105 4.940.002 4.940.005 4.945.104 4.945.103 4.940.103
WITG4.940.105EA 38.2 EUR
WITG4.940.105 WITG4.940.002 WITG4.940.005 WITG4.945.104 WITG4.945.103 WITG4.940.103
Centrifuge tubes, round bottom
Putket Sentrifugiputket
DURAN®, borosilicate glass.
  • Heavy wall
  • Very good chemical resistance
  • Very good temperature resistance
Round bottom, rimless.