Weighing bottles, glass
Toimittaja: witeg Labortechnik
2.872.005
WITG2.872.005EA 31.8 EUR
WITG2.872.005
Weighing bottles, glass
Pullot Punnituslasit
Made of borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Permanent graduations and measures
Low form.