Separating funnels, Squibb
Toimittaja: witeg Labortechnik
2.649.125 2.649.150
WITG2.649.125EA 126 EUR
WITG2.649.125 WITG2.649.150
Separating funnels, Squibb
Suppilot Erotussuppilot
Separatory funnels, Squibb, made of borosilicate glass 3.3, stopcock with economy PTFE-needle valve.
  • Easy to read graduation
  • Very good chemical resistance
  • Very good temperature resistance