Büchner funnels
Toimittaja: witeg Labortechnik
2.367.124
WITG2.367.124EA 95.8 EUR
WITG2.367.124
Büchner funnels
Suppilot Suodatussuppilot Büchner-suppilot
Funnels, made of borosilicate glass 3.3, with sintered disc, for convenient filtration of solutions.
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance