Dropping funnels, cylindrical, with pressure balance
Toimittaja: witeg Labortechnik
1.869.025 1.869.050 1.869.100 1.869.250
WITG1.869.025EA 363 EUR
WITG1.869.025 WITG1.869.050 WITG1.869.100 WITG1.869.250
Dropping funnels, cylindrical, with pressure balance
Suppilot Tippasuppilot, NS-hios
Dropping funnels, cylindrical, made of borosilicate glass, with needle valve stopcock with PTFE-needle valve.
  • Graduated
  • With NS cone and socket
  • Very good chemical and temperature resistance
With screw-thread retaining nut.