Stirrer guides
Toimittaja: witeg Labortechnik
1.460.029 1.470.029 1.468.004 1.468.029 1.475.029
WITG1.460.029EA 134 EUR
WITG1.460.029 WITG1.470.029 WITG1.468.004 WITG1.468.029 WITG1.475.029
Stirrer guides
Sekoittajat Sekoitustyökalut
Stirrer guides are of different types.
With fused-on stirrer bearing.
GTS-vacuum-stirrer, with PTFE-insert and interchangeable PTFE-vacuum-seal, screw cap GL 25.
With PP-insert, 8 mm Ø and screw cap GL 25, made of melamine.
With cooling jacket and PTFE-insert, with interchangeable PTFE-vacuum seal and screw cap GL 25, made of melamine.