Reaction vessels, cylindrical, with flat flange
Toimittaja: witeg Labortechnik
0.900.100
WITG0.900.100EA 95.6 EUR
WITG0.900.100
Reaction vessels, cylindrical, with flat flange
Reaktioastiat Reaktioastiat, sylinterin muotoiset
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With graduation, flange with groove and round bottom.