WHEATON® COMPLETEPACK Vials with Stoppers and Seals, Clear Glass
Toimittaja: DWK Life Sciences
W991000-CP W991001-CP W991002-CP W991003-CP W991004-CP W991005-CP W991006-CP W991007-CP W991008-CP W991009-CP W991010-CP W991011-CP W991012-CP W991013-CP W991014-CP W991015-CP W991016-CP W991017-CP
WHEAW991000-CPEA 5380 EUR
WHEAW991000-CP WHEAW991001-CP WHEAW991002-CP WHEAW991003-CP WHEAW991004-CP WHEAW991005-CP WHEAW991006-CP WHEAW991007-CP WHEAW991008-CP WHEAW991009-CP WHEAW991010-CP WHEAW991011-CP WHEAW991012-CP WHEAW991013-CP WHEAW991014-CP WHEAW991015-CP WHEAW991016-CP WHEAW991017-CP
WHEATON® COMPLETEPACK Vials with Stoppers and Seals, Clear Glass
Näytevialit ja pienet näyteastiat
Complete range of clear crimp top vials, rubber stoppers, and seals that are sterilised and ready-to-use.
  • Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Complete manufacturing and certificate of analysis traceability
  • Container Closure Integrity (CCI) validation
Ready-to-use, just-in-time, minimum 180 day shelf life from time of shipment.
Sertifikaatit: USP 788 / EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers), USP 85 /EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers) and lot certified to SAL 10⁻More Product Information