WHEATON® CompletePAK® Sterile Components, Vials Only
Toimittaja: DWK Life Sciences
W987000-S W987001-S W987002-S W987000A-S W987001A-S W987002A-S
WHEAW987000-SEA 2170 EUR
WHEAW987000-S WHEAW987001-S WHEAW987002-S WHEAW987000A-S WHEAW987001A-S WHEAW987002A-S
WHEATON® CompletePAK® Sterile Components, Vials Only
Näytevialit ja pienet näyteastiat
Clear or amber glass crimp finish vials sterilised, ready-to-use (RTU), complete manufacturing/certificate of analysis traceability included.
  • Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Elastomeric closures used in parenteral preparations
  • Complete manufacturing and certificate of analysis traceability
  • Ready-to-use, just-in-time, minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
  • Available in clear or amber glass for photosensitive contents
Sertifikaatit: USP 788 / EP 2.9.19More Product Information