Screw caps, WHEATON®
Toimittaja: DWK Life Sciences
240209 240218 240280 240408 240409 240680 W240540 240415 240416 240418 240480 240206 240208 240215 240216 W240406 W240517 240481 240463 239853 W240508 W240509 240281 W240518 240419 W240516 240080 240119 240121 240181 240081 240223 240228 239253 239249 239251 239255 239257 239259 239260 239250 239451 240264 240319 240321 240330 240381 240414 240421 240269 W239302 W240820 W240824 W240825 W240822 W239299 W239300 W239301 W239304 W239303 W239298 W240823
548-2152EA 183 EUR
548-2152 548-2155 WHEA240280 548-2030 548-2031 WHEA240680 218-2147 548-2032 548-2033 548-2034 548-2035 548-2150 548-2151 548-2153 548-2154 WHEAW240406 WHEAW240517 WHEA240481 WHEA240463 WHEA239853 WHEAW240508 WHEAW240509 WHEA240281 WHEAW240518 WHEA240419 WHEAW240516 WHEA240080 WHEA240119 WHEA240121 WHEA240181 WHEA240081 WHEA240223 WHEA240228 WHEA239253 WHEA239249 WHEA239251 WHEA239255 WHEA239257 WHEA239259 WHEA239260 WHEA239250 WHEA239451 WHEA240264 WHEA240319 WHEA240321 WHEA240330 WHEA240381 WHEA240414 WHEA240421 WHEA240269 WHEAW239302 WHEAW240820 WHEAW240824 WHEAW240825 WHEAW240822 WHEAW239299 WHEAW239300 WHEAW239301 WHEAW239304 WHEAW239303 WHEAW239298 WHEAW240823
Screw caps, WHEATON®
Tulpat ja korkit Pullojen sulkimet
Rubber lined caps feature a non-toxic white styrene-butadiene rubber liner ideal for cell culture work.
  • Aluminum foil faced pulp board liners can be used with organic solvents. Good resistance to hydrocarbons, oils, ketones and alcohols Not good for acids or alkalis.
Use low density polyethylene (LDPE) lined caps with distilled water, analytical standards and reagents.
PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product.
Bonded PTFE faced silicone liner ideal for storage ofMore Product Information